missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Chloride Solution, For Sulfide, 6N, LabChem™

Supplier:  LabChem LC108201

 View more versions of this product

Catalog No. LC108201


Explore available promotions

Description

Description

Specifications

Specifications

Aluminum Chloride Solution
For Sulfide
50,50
AlCl3·6H2O
UN2581
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminium(3+) hexahydrate trichloride
22377415
241.43
Poly Bottle
500 mL
Colorless to Yellow
7784-13-6,7732-18-5
AlCl3H12O6
MFCD00149134
6N
Soluble in water
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
241.42
CHEBI:74779
Passes Test
Hydrogen chloride
Liquid

Chemical Identifiers

7784-13-6
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.

GHS P Statement
Avoid breathing mist.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions