Supertek™ Aluminum Density Set DFS Item
Demonstrate that although the mass and volume for each example are different, the density and specific gravity will remain the same. Includes 12 aluminum cylinders of various lengths in a wood storage block.
Supplier:  Supertek™ PH100483A
Catalog No. S012343
Explore available promotions
Description
Description
  • Demonstrate that although the mass and volume for each example are different, the density and specific gravity will remain the same.
  • Includes 12 aluminum cylinders (0.5 in. dia.) of various lengths.
  • Includes wood storage block.
Specifications
Specifications
Density Set