missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum fluoride, anhydrous, 99+%, Thermo Scientific Chemicals

$44.74 - $662.90

Chemical Identifiers

CAS 7784-18-1
MDL Number MFCD00003426
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1149703
View Documents
Thermo Scientific Chemicals
01149703
1 g
Each for $44.74
 
AA1149709
View Documents
Thermo Scientific Chemicals
01149709
10 g
Each for $184.44
 
AA1149718
View Documents
Thermo Scientific Chemicals
01149718
50 g
Each for $662.90
 
Description

Description

In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactionsAluminum fluoride is used in the smelting process to lower the melting point of electrolytes, production of aluminum silicates, refining of aluminum scarp, production of specialty refractory products, and in glass industry as filler. It is used as flux ingredient to remove magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions
Specifications

Chemical Identifiers

7784-18-1
MFCD00003426

Specifications

1291°C (sublimation)
Powder
MFCD00003426
Practically insoluble in water; Sparingly soluble in acids and alkalies.
≥99%
2.882 g/mL
7784-18-1
1 g
14,339
83.98
Hygroscopic
Aluminum fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

EINECSNumber : 232-051-1

RTECSNumber : BD0725000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only