missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Fluoride, Powder, 60%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A109050KGBL

 View more versions of this product

Catalog No. 18608513

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

15098-87-0
50 kg
Reagent
1
60%
Drum

Chemical Identifiers

15098-87-0
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.