missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum fluoride trihydrate, 97%, Thermo Scientific Chemicals

$89.11 - $724.99

Chemical Identifiers

CAS 15098-87-0
Molecular Formula AlF3H6O3
Molecular Weight (g/mol) 138.022
MDL Number MFCD00003426
InChI Key BKLZIAYVINRQEJ-UHFFFAOYSA-K
Synonym aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
PubChem CID 22002992
IUPAC Name aluminum;trifluoride;trihydrate
SMILES O.O.O.[F-].[F-].[F-].[Al+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1149622
View Documents
Thermo Scientific Chemicals
01149622
100 g
Each for $89.11
 
AA1149636
View Documents
Thermo Scientific Chemicals
01149636
500 g
Each for $253.84
 
AA11496A3
View Documents
Thermo Scientific Chemicals
011496A3
2 kg
Each for $724.99
 
Description

Description

Aluminum fluoride trihydrate is used as an additive for the production of aluminum by electrolysis. It is also used to inhibit fermentation and in the preparation of low index films. It serves as a flux ingredient for the removal of magnesium in the refining of aluminum scrap. Further, it is used in the manufacture of aluminum silicates, ceramics and glass. In addition, it acts as a catalyst for organic synthesis and filler in glass industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15098-87-0
138.022
BKLZIAYVINRQEJ-UHFFFAOYSA-K
22002992
O.O.O.[F-].[F-].[F-].[Al+3]
AlF3H6O3
MFCD00003426
aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
aluminum;trifluoride;trihydrate

Specifications

250°C (in -3H2O)
White
100 g
MFCD00003426
aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
BKLZIAYVINRQEJ-UHFFFAOYSA-K
aluminum;trifluoride;trihydrate
22002992
97%
2.17 g/mL
15098-87-0
Powder
AlF3H6O3
14,339
Slightly soluble in water,acids and alkalis. Insoluble in alcohol and acetone.
O.O.O.[F-].[F-].[F-].[Al+3]
138.022
138.02 (83.98 Anhydrous)
Odorless
Aluminum fluoride trihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H320-H335

EINECSNumber : 232-051-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only