Learn More
Aluminum nitrate nonahydrate, 98%, Thermo Scientific Chemicals
$82.23 - $681.23
Chemical Identifiers
CAS | 7784-27-2 |
---|---|
Molecular Formula | AlH18N3O18 |
Molecular Weight (g/mol) | 375.129 |
MDL Number | MFCD00003419 |
InChI Key | SWCIQHXIXUMHKA-UHFFFAOYSA-N |
Synonym | aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic |
PubChem CID | 24567 |
IUPAC Name | aluminum;trinitrate;nonahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1236014
|
Thermo Scientific Chemicals
01236014 |
25 g |
N/A
|
|
|||||
AA1236030
|
Thermo Scientific Chemicals
01236030 |
250 g |
N/A
|
|
|||||
AA12360A1
|
Thermo Scientific Chemicals
012360A1 |
1 kg |
N/A
|
|
|||||
AA12360A7
|
Thermo Scientific Chemicals
012360A7 |
5 kg |
N/A
|
|
|||||
Description
It finds use in tanning leather, antiperspirant, corrosion protection, extraction of uranium, nitrating agent. It is an effective dopant for producing highly conductive thin film by utilizing the sol gel method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-27-2 | |
375.129 | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
24567 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
AlH18N3O18 | |
MFCD00003419 | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
aluminum;trinitrate;nonahydrate |
Specifications
73.5°C (135° (decomposition)) | |
White | |
25 g | |
AlH18N3O18 | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate | |
24567 | |
Odorless | |
White crystalline |
7784-27-2 | |
Crystals | |
98% | |
MFCD00003419 | |
Soluble in water,ethanol,glycerol and methanol. | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
375.129 | |
375.13 (212.99 Anhydrous) | |
Hygroscopic | |
Aluminum nitrate nonahydrate, nonahydrate |
Safety and Handling
P280-P305+P351+P338-P310
H318
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 236-751-8
RTECSNumber : BD1050000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only