Learn More
Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals
$55.74 - $1735.07
Chemical Identifiers
CAS | 814-87-9 |
---|---|
Molecular Formula | C6Al2O12 |
Molecular Weight (g/mol) | 318.02 |
MDL Number | MFCD00050791 |
InChI Key | ZCLVNIZJEKLGFA-UHFFFAOYSA-H |
Synonym | aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate |
PubChem CID | 93561 |
IUPAC Name | dialuminum;oxalate |
SMILES | [Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4414909
|
Thermo Scientific Chemicals
04414909 |
10 g |
Each for $55.74
|
|
|||||
AA4414918
|
Thermo Scientific Chemicals
04414918 |
50 g |
Each for $159.15
|
|
|||||
AA4414930
|
Thermo Scientific Chemicals
04414930 |
250 g |
Each for $596.55
|
|
|||||
AA44149A1
|
Thermo Scientific Chemicals
044149A1 |
1 kg |
Each for $1,735.07
|
|
|||||
Description
Aluminum oxalate is used as a mordant for printing textiles and dyeing cotton. It finds use as a laboratory reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
814-87-9 | |
318.02 | |
ZCLVNIZJEKLGFA-UHFFFAOYSA-H | |
93561 | |
[Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
C6Al2O12 | |
MFCD00050791 | |
aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate | |
dialuminum;oxalate |
Specifications
814-87-9 | |
C6Al2O12 | |
MFCD00050791 | |
14,355 | |
Insoluble in water. | |
[Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O | |
318.02 | |
318.02 (Anhydrous) | |
Chunks and/or Powder |
White | |
Al2(C2O4)3·xH2O | |
10 g | |
aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate | |
ZCLVNIZJEKLGFA-UHFFFAOYSA-H | |
dialuminum;oxalate | |
93561 | |
99% | |
Aluminum oxalate hydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
EINECSNumber : 212-407-2
RTECSNumber : BD0516250
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only