missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum oxide, alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04257122

 View more versions of this product

Catalog No. AA4257122


Explore available promotions

Description

Description

Alumium oxide is widely used in a variety of plastics, rubber, ceramics, refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire, yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamp as far-infrared emission and thermal insulation materials. It is useful for the C single crystal detectors. It plays an important role for the hydrogenation of aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2980°C
2045°C
1344-28-1
100 g
Al2O3
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
Odorless
Aluminum oxide
White
Powder
99.9% (Metals basis)
MFCD00003424
Slowly soluble in aqueous alkali solutions-forming hydroxides. Insoluble in water and in non-polar organic solvents.
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
1.765

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only