missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Aluminum Sign: EAR PLUGS MUST BE WORN IN THIS AREA. FAILURE MAY RESULT IN HEARING DAMAGE.
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$19.10 - $34.50
Specifications
Material | Aluminum |
---|---|
Shape | Rectangular |
Displayed Text | EAR PLUGS MUST BE WORN IN THIS AREA. FAILURE MAY RESULT IN HEARING DAMAGE. |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
19-111-747
|
Brady™
144296 |
10 x 7 in. |
Each for $19.10
|
|
|||||
19-111-742
|
Brady™
144291 |
14 x 10 in. |
Each for $34.50
|
|
|||||
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Aluminum | |
Rectangular | |
EAR PLUGS MUST BE WORN IN THIS AREA. FAILURE MAY RESULT IN HEARING DAMAGE. |