missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum Sulfate Hydrate ACS MP Biomedicals
Supplier: MP Biomedicals Inc 0219140380
Specifications
Aluminum Sulfate Hydrate | |
17927-65-0 | |
98 to 102% | |
MFCD00149136 | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
dialuminium(3+) trisulfate | |
22377415 | |
342.15 (Anhydrous) |
Hydrate | |
100 g | |
Al2O12S3 | |
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
342.13 | |
CHEBI:74779 | |
ACS Reagent |
Chemical Identifiers
17927-65-0 | |
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
CHEBI:74779 |
MFCD00149136 | |
22377415 |
Safety and Handling
Recommended Storage : Store at room temperature.