missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Aluminum Sulfate Hydrate (Crystalline/Certified ACS), Fisher Chemical
Supplier: Fisher Chemical FLA613500
Specifications
Aluminum Sulfate Hydrate | |
White | |
3 to 4 | |
500 g | |
Al2H36O30S3 | |
MFCD00149136 | |
AMVQGJHFDJVOOB-UHFFFAOYSA-H | |
dialuminum;trisulfate;octadecahydrate | |
22377415 | |
666.42 (for 18-hydrate) | |
Certified ACS | |
Poly Bottle |
86°C | |
7784-31-8 | |
Powder/Solid | |
98.0 to 102.0 % | |
Al2(SO4)3·nH20 | |
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] | |
666.401 | |
CHEBI:74779 | |
98.0 to 102.0% | |
Pass Test |
Chemical Identifiers
7784-31-8 | |
666.401 | |
AMVQGJHFDJVOOB-UHFFFAOYSA-H | |
22377415 | |
dialuminum;trisulfate;octadecahydrate |
Al2H36O30S3 | |
MFCD00149136 | |
aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
CHEBI:74779 | |
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
Safety and Handling
