Fisherbrand™ Amber Boston Round Glass Bottles with Black Phenolic Caps
Amber Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with black phenolic caps attached to maintain cleanliness
$66.52 - $599.48
Specifications
Color | Amber |
---|---|
Material | Glass (Type 3) |
Closure Color | Black |
Closure Included | Yes |
Mouth | Narrow |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Liner | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Liner | Quantity | Price | Quantity | |||
FB02912339
|
N/A | 30 mL | Polyethylene Cone | 48 |
Case of 48 Each for $112.52
|
|
|||
FB02911898
|
N/A | 60 mL | Polyethylene Cone | 288 |
Case of 288 Each for $599.48
|
|
|||
FB02911734
|
N/A | 60 mL | Polyethylene Cone | 24 |
Case of 24 Each for $66.52
|
|
|||
FB02911735
|
N/A | 120 mL | Polyethylene Cone | 24 |
Case of 24 Each for $84.40
|
|
|||
FB02911895
|
N/A | 120 mL | Polyethylene Cone | 160 |
Case of 160 Each for $435.60
|
|
|||
FB02912341
|
N/A | 120 mL | Rubber | 24 |
Case of 24 Each for $91.24
|
|
|||
FB02912342
|
N/A | 240 mL | Rubber | 12 |
Case of 12 Each for $70.84
|
|
|||
FB02911900
|
N/A | 240 mL | Polyethylene Cone | 108 |
Case of 108 Each for $393.44
|
|
|||
FB02911743
|
N/A | 480 mL | Polyethylene Cone | 12 |
Case of 12 Each for $71.52
|
|
|||
FB02911878
|
N/A | 480 mL | Polyethylene Cone | 60 |
Case of 60 Each for $297.84
|
|
|||
FB02912344
|
N/A | 950 mL | Rubber | 12 |
Case of 12 Each for $117.88
|
|
|||
FB02911744
|
N/A | 950 mL | Polyethylene Cone | 12 |
Case of 12 Each for $112.84
|
|
|||
Description
Amber Boston round Type III soda-lime glass is ideal for solvent, chemical or sample storage. Packaged in convenience packs with black phenolic caps attached to maintain cleanliness
- Narrow screw neck opening, curved shoulders, and a round body
- Glass offers minimal chemical interference with contents
- Amber color absorbs all wavelengths in the ultraviolet spectrum
- Black Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier
- Liners mold themselves around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
- Ideal for everything from liquids, to soaps, to macro-invertebrates.
Specifications
Amber | |
Black | |
Narrow | |
Phenolic |
Glass (Type 3) | |
Yes | |
Boston Round Bottle |