Fisherbrand™ Amber Boston Round Glass Bottles without Caps
Type III soda-lime glass is ideal for solvent, chemical or sample storage
 View more versions of this product
Catalog No. FB02911905
Explore available promotions
Description
Description
  • Features narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
  • Ensure integrity for long term storage and protection for UV-sensitive contents
Specifications
Specifications
240 mL
Amber
13 mm
108
No
136 mm
24-400 mm
8 oz.
21 mm
Glass (Type 3)
24-400
Narrow
Boston Round Bottle
60 mm