missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Amber Reagent Bottle with Screw Cap
High quality, borosilicate glass flask. Pouring ring for drip free pouring and clean, safe working.
Supplier: Eisco™ CH0165A
Description
- Reagent bottle made of high quality, amber colored Borosilicate 3.3 glass.
- Bottle features white graduation lines.
- As per DIN ISO 4796-1 standard.
- Equipped with an orange, GL 45 screw cap and features a drip free pouring ring for cleaner, safer transfers of liquids and powders.
- Both screw cap and pouring ring are autoclavable.
- Bottle features superior chemical durability and low temperature gradient - can withstand high temperatures and thermal shocks.
Specifications
100 mL | |
3.3 oz. | |
For storing chemicals, liquids or powders | |
1 Ea. | |
Orange | |
Yes | |
Reusable | |
Narrow | |
11.7 cm | |
4.6 in. | |
Polypropylene | |
Screw Cap |
100 mL | |
Amber | |
Borosilicate Glass | |
Non-sterile | |
10mL | |
Autoclavable | |
Yes | |
Cylindrical | |
Reagent Bottle | |
Blank | |
GL 45 |