missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Amber Reagent Bottle with Screw Cap

High quality, borosilicate glass flask. Pouring ring for drip free pouring and clean, safe working.

Supplier:  Eisco™ CH0165E

 View more versions of this product

Catalog No. S27990


Explore available promotions

Description

Description

  • Reagent bottle made of high quality, amber colored Borosilicate 3.3 glass.
  • Bottle features white graduation lines.
  • As per DIN ISO 4796-1 standard.
  • Equipped with an orange, GL 45 screw cap and features a drip free pouring ring for cleaner, safer transfers of liquids and powders.
  • Both screw cap and pouring ring are autoclavable.
  • Bottle features superior chemical durability and low temperature gradient - can withstand high temperatures and thermal shocks.
Specifications

Specifications

2000 mL
68 oz.
For storing chemicals, liquids or powders
1 Ea.
Orange
Yes
Reusable
Narrow
260.35 mm
10.25 in.
GL45
Screw Cap
2000 mL
Amber
Borosilicate Glass
Non-sterile
250mL
Autoclavable
Yes
Cylindrical
Reagent Bottle
Blank
25.4 mm
Videos
SDS
Documents

Documents

Promotions

Promotions