Learn More
Amberlite™ IRC-120(H), ion exchange resin, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L142850B
Description
Amberlite™ IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmberlite|r IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Amberlite™ IR-120(H) | |
78922-04-0 | |
MFCD00132707 | |
14,382 | |
3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid | |
APBOVLPLJFJSRI-UHFFFAOYSA-N | |
3-[(3-chlorophenyl)sulfonylamino]benzoic acid | |
8190984 |
ion exchange resin | |
C13H10ClNO4S | |
1.28 | |
1000 g | |
Insoluble in water. | |
C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O | |
311.736 |
Chemical Identifiers
78922-04-0 | |
311.736 | |
APBOVLPLJFJSRI-UHFFFAOYSA-N | |
8190984 | |
C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O |
C13H10ClNO4S | |
MFCD00132707 | |
3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid | |
3-[(3-chlorophenyl)sulfonylamino]benzoic acid |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only