missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amberlite™ IRC120 Na ion exchange resin, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1519922

 View more versions of this product

Catalog No. AAL1519922


Explore available promotions

Description

Description

Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

Solubility
Insoluble in water.

Notes
Water softening, dealkalinization, deionization
Specifications

Specifications

Amberlite™ IR-120(Na) ion exchange resin
C13H10ClNO4S
14,382
3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
APBOVLPLJFJSRI-UHFFFAOYSA-N
3-[(3-chlorophenyl)sulfonylamino]benzoic acid
8190984
78922-04-0
MFCD00132707
100 g
Insoluble in water.
C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O
311.736

Chemical Identifiers

78922-04-0
311.736
APBOVLPLJFJSRI-UHFFFAOYSA-N
8190984
C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O
C13H10ClNO4S
MFCD00132707
3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
3-[(3-chlorophenyl)sulfonylamino]benzoic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only