missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amberlite™ MAC-3 H, Hydrogen Form, MilliporeSigma™ Supelco™
Dowex MAC-3 contains carboxylic acid as functional group and is selective for Cu and Fe. The resin has larger exchange capacity and large particle size.
$106.43 - $337.66
Specifications
CAS | 9052-45-3 |
---|---|
Type | Organic Standard |
Product Line | Amberlite |
MDL Number | MFCD03098973 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow