Learn More
Amberlyst™ 15(H), ion exchange resin, Thermo Scientific Chemicals
Strongly acidic macroreticular resin
$46.96 - $428.68
Chemical Identifiers
CAS | 39389-20-3 |
---|---|
Molecular Formula | C18H18O3S |
Molecular Weight (g/mol) | 314.399 |
MDL Number | MFCD00145841 |
InChI Key | SIWVGXQOXWGJCI-UHFFFAOYSA-N |
Synonym | amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid |
PubChem CID | 170197 |
IUPAC Name | 1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid |
SMILES | C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA8907918
|
Thermo Scientific Chemicals
08907918 |
50 g |
Each for $46.96
|
|
|||||
AA8907930
|
Thermo Scientific Chemicals
08907930 |
250 g |
Each for $131.53
|
|
|||||
AA89079A1
|
Thermo Scientific Chemicals
089079A1 |
1 kg |
Each for $428.68
|
|
|||||
Description
Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
39389-20-3 | |
314.399 | |
SIWVGXQOXWGJCI-UHFFFAOYSA-N | |
170197 | |
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O |
C18H18O3S | |
MFCD00145841 | |
amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid | |
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid |
Specifications
Pale brown beads | |
Odorless | |
MFCD00145841 | |
50 g | |
Insoluble in water. | |
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O | |
314.399 | |
Amberlyst™ 15(H) |
39389-20-3 | |
C18H18O3S | |
Brown | |
amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid | |
SIWVGXQOXWGJCI-UHFFFAOYSA-N | |
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid | |
170197 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only