missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Magnetic-Ring Stand (96 well) DFS Item

The 96-well Magnetic-Ring Stand is designed for paramagnetic bead precipitation from standard 96-well U-bottom microplates.

Supplier:  Invitrogen™ AM10050

Catalog No. AM10050


Explore available promotions

Description

Description

  • Stand has plastic base in standard SBS 96-well microtiter plate footprint
  • Uses 96 extremely strong permanent magnetic rings for fast, easy magnetic separation
  • Paramagnetic beads in U-bottom plate pellet in donut shape approximately 6mm in diameter, allowing easy removal of supernatant by robot or multichannel pipettor
  • Stand sits flat on lab bench or in microtiter plate adapter on robotic workstations

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature.
Room Temperature
1 Each
Magnetic Stand
Ambion™
Magnetic Stand
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.