missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Cells-to-CT™ Bulk Lysis Reagents DFS Item

Cells-to-CT™ Bulk Lysis Reagents are components of the TaqMan™ Gene Expression Cells-to-CT Kit, made available as a separate purchase, in larger volumes than provided with the kit.

Supplier:  Invitrogen™ 4391851C

Catalog No. 4391851C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Includes: Lysis Solution (124mL), Stop Solution (12.5mL), DNase (1.25mL)

Description

Description

Cells-to-CT™ Bulk Lysis Reagents are components of the TaqMan™ Gene Expression Cells-to-CT Kit, made available as a separate purchase, in larger volumes than provided with the kit. The reagents included are: Lysis Solution, Stop Solution, and DNase. The TaqMan Gene Expression Cells-to-CT Kit enables researchers to easily perform expression analysis using cultured cells without purification of RNA.

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

This kit contains the following:
• Lysis Solution (124 mL): Store at 4°C
• Stop Solution (12.5 mL): Store at -20°C
• DNase (1.25 mL): Store at -20°C

2500 reactions
Cell Lysis Buffer
Dry Ice
Fewer resources used and less waste, Less hazardous
Cells-to-CT™
2,500 reactions
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.