Invitrogen™ DEPC-Treated Water
Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC.
Supplier: Invitrogen™ AM9906
Description
Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Specifications
6 to 8 | |
Bottle | |
10 x 50 mL | |
Room Temperature |
Molecular Biology | |
DEPC-Treated | |
Autoclaved, Membrane-Filtered | |
Ambion™ |
For Research Use Only. Not for use in diagnostic procedures.