Learn More
Invitrogen™ EDTA (0.5 M), pH 8.0, RNase-free DFS Item
Invitrogen Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use.
Supplier:  Invitrogen™ AM9260G
 View more versions of this product
Catalog No. AM9260G
Only null left
Explore available promotions
Description
Description
Invitrogen Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Specifications
Specifications
EDTA
0.5 M
8
Molecular Biology Grade
Room Temperature
Store at room temperature.
Bottle
Ambion
100 mL
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.