missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Elution Solution, Nucleic Acid Purification
The Elution Solution is used in the purification of total RNA from cells, primary cell isolates, animal and plant tissues and tumors.
Supplier: Invitrogen™ 4305893
Description
The Elution Solution is used in the purification of total RNA from cells, primary cell isolates, animal and plant tissues and tumors.
- For use with Total RNA Lysis Solution and Nucleic Acid Purification
- Size: 1000 mL
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
Nucleic Acid Purification | |
1000 mL | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.