Learn More
Invitrogen™ FirstChoice™ RLM-RACE Kit DFS Item
Designed to amplify cDNA only from full-length, capped mRNA, usually producing a single band after PCR
Supplier:  Invitrogen™ AM1700M
 View more versions of this product
Catalog No. AM1700M
Only null left
Explore available promotions
Description
Description
FastAP™ Thermosensitive Alkaline Phosphatase, 10X FastAP Buffer, Tobacco Acid Pyrophosphatase, 10X TAP Buffer, T4 RNA Ligase, 10X T4 RNA Ligase Buffer, M-MLV Reverse Transcriptase, RNase Inhibitor, 10X RT Buffer, 2.5 mM dNTP Mix, Mouse Thymus RNA (1 mg/ml), Random Decamers, 5' RACE Adapter, 3' RACE Adapter, 5' RACE Outer Primer, 5' RACE Inner Primer, 3' RACE Outer Primer, 3' RACE Inner Primer, 5' RACE Outer Control Primer, 5' RACE Inner Control Primer, 5' PCR Control Primer, 3' RACE Control Primer and Ammonium Acetate Stop Solution should be stored at -20°C. Nuclease-free Water may be stored at any temperature.
The FirstChoice™ RLM-RACE Kit is designed to amplify cDNA from full-length, capped mRNA through RNA Ligase Mediated Rapid Amplification of cDNA Ends (RLM-RACE). The kit ensures the amplification of only full-length transcripts by eliminating ribosomal RNA, fragmented mRNA, tRNA, and contaminating genomic DNA during the amplification process. This kit is a major improvement over the basic rapid amplification of cDNA ends (RACE) protocol.
Note: calf intestinal phosphatase (CIP) had been replaced with FastAP Thermosensitive Alkaline Phosphatase adjusted protocol eliminate the need to use phenol/chloroform extraction and ethanol precipitation. If chosen, phenol/chloroform step can be used with updated kit after verifying reaction volumes.
Features of the FirstChoice RLM-RACE Kit include:
  • Selects 5' and/or 3' ends of full-length, capped mRNA
  • Optimized to ensure detection of even the rare mRNA, yielding specific product
  • Allows to finish experiment in less than a day
Selects full-length mRNA—no rRNA, tRNA or degraded mRNA
In the FirstChoice RLM-RACE procedure, full-length mRNAs are selected by treating total or poly(A) RNA with FastAP Thermosensitive Alkaline Phosphatase to remove the 5'-phosphate from all molecules which contain free 5'-phosphates (ribosomal RNA, fragmented mRNA, tRNA, and contaminating genomic DNA). Full-length mRNAs are unaffected. The RNA is then treated with tobacco acid pyrophosphatase (TAP) to remove the cap structure from the full-length mRNA leaving a 5'-monophosphate. A synthetic RNA adapter is ligated to the RNA population—only molecules containing a 5'-phosphate, the uncapped, full-length mRNAs, will accept the adapter. Random-primed, reverse transcription reactions and nested PCR are then performed to amplify the 5'-end of your specific transcript.
From RNA to PCR in less than a day
Each step of the FirstChoice RLM-RACE procedure has been optimized, so you can complete all the enzymatic reactions and even start PCR in no more than 5 hours. The FirstChoice RLM-RACE Kit contains reagents and enzymes to produce 6 RLM-RACE-ready cDNA preparations, primers and nested RACE adapter primers for 100 PCR reactions. Kit includes reverse transcription reagents and 3' RACE Adapter and Primers for 3' RACE. Each kit contains control RNA and primers to test the kit's performance, protocol described in User Guide utilizing Platinum™ II Hot-Start PCR Master Mix.
Specifications
Specifications
RLM-RACE Kit
•FastAP™ Thermosensitive Alkaline Phosphatase
•10X FastAP Buffer, Tobacco Acid Pyrophosphatase
•10X TAP Buffer, T4 RNA Ligase
•10X T4 RNA Ligase Buffer
•M-MLV Reverse Transcriptase
•RNase Inhibitor
•10X RT Buffer
•2.5 mM dNTP Mix
•Mouse Thymus RNA (1 mg/ml)
•Random Decamers
•5' RACE Adapter
•3' RACE Adapter
•5' RACE Outer Primer
•5' RACE Inner Primer
•3' RACE Outer Primer
•3' RACE Inner Primer
•5' RACE Outer Control Primer
•5' RACE Inner Control Primer
•5' PCR Control Primer
•3' RACE Control Primer
•Ammonium Acetate Stop Solution
•Components should be stored at -20°C. Nuclease-free Water may be stored at any temperature.
42°C
Kit
cDNA Libraries & Library Construction
Kit with manual, FastAP™ Thermosensitive Alkaline Phosphatase, 10X FastAP Buffer, Tobacco Acid Pyrophosphatase, 10X TAP Buffer, T4 RNA Ligase, 10X T4 RNA Ligase Buffer, M-MLV Reverse Transcriptase, RNase Inhibitor, 10X RT Buffer, 2.5 mM dNTP Mix, Mouse Thymus RNA (1 mg/ml), Random Decamers, 5’ RACE Adapter, 3’ RACE Adapter, 5’ RACE Outer Primer, 5’ RACE Inner Primer, 3’ RACE Outer Primer, 3’ RACE Inner Primer, 5’ RACE Outer Control Primer, 5’ RACE Inner Control Primer, 5’ PCR Control Primer, 3’ RACE Control Primer and Ammonium Acetate Stop Solution, Nuclease-free Water
cDNA
M-MLV
Ambion, FirstChoice
1 Kit with manual
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.