missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Magnetic Stand-96 DFS Item

The stand has clear polyacryl base in standard SBS 96-well microplate footprint and uses 24 extremely strong permanent magnets for fast, easy magnetic separation in little as 30 seconds.

Supplier:  Invitrogen™ AM10027

Catalog No. AM10027


Explore available promotions

Description

Description

The stand has clear polyacryl base in standard SBS 96-well microplate footprint and uses 24 extremely strong permanent magnets for fast, easy magnetic separation in little as 30 seconds. The stand sits flat on lab bench or microplate adapter on robotic workstations.

For use with MagMAX™-96 magnetic bead-based RNA isolation kits, MagMAX™ for Stabilized Blood Tubes RNA Isolation Kits, MagMAX™ FFPE Nucleic Acid Isolation Kits, and the MessageAmp™ Premier RNA Amplification Kit

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature.
Room Temperature
1 Ea.
Magnetic Stand
Ambion™
Magnetic Stand
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.