Invitrogen™ Nuclease-Free Water (not DEPC-Treated) DFS Item
Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving.
Supplier:  Invitrogen™ AM9939
 View more versions of this product
Catalog No. AM9939
Explore available promotions
Description
Description
Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving. This nuclease-free water is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Using Nuclease-free Water
Because this water is not DEPC-treated, it is recommended for applications for which residual DEPC might prove detrimental (e.g., oocyte microinjection).
Specifications
Specifications
6 to 8
Bottle
5 x 100 mL
Room Temperature
Molecular Biology
Not DEPC-Treated
Autoclaved, Membrane-Filtered
Ambion™
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.