Invitrogen™ Ambion™ RNase A, affinity purified, 1 mg/mL DFS Item
Rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity
Supplier:  Invitrogen™ AM2270
 View more versions of this product
Catalog No. AM2270
Explore available promotions
Description
Description
Ambion RNase A is endonuclease that specifically cleaves 3' of U and C residues. Affinity Purified RNase A is intended for critical applications when absence of DNase and other nonspecific nuclease activities is essential.
  • Supplied in one tube containing 200μg (1mg/mL)
  • Preparation contains RNase B, a carbohydrate isoform of RNase A
DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification
Specifications
Specifications
1 mg/mL
Dry Ice
200 μg
Ambion™
-20°C
RNase
RNase A
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.