Invitrogen™ RNase Cocktail™ Enzyme Mix DFS Item
Endonuclease-free, exonuclease-free, protease-free
$173.00 - $512.00
Specifications
Shipping Condition Dry Ice
Enzyme RNase
Product Type RNAse Cocktail Enzyme Mix
Product Line Ambion™
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AM2286 DFS Item
View Documents
Invitrogen™
AM2286
1 mL
Each for $173.00
AM2288 DFS Item
View Documents
Invitrogen™
AM2288
5 x 1 mL
Each for $512.00
Description
Description
RNase Cocktail™ is mixture of two highly purified ribonucleases, RNase A (500U/mL) and RNase T1 (20,000U/mL) and is free of DNase and nicking activities. Digestion of RNA with RNase A alone leaves fragments of RNA which are large enough to be visible on agarose gels and precipitate in ethanol. RNase A cuts after C and U residues, and RNase T1 cuts after G residues. Consequently, mixture of both enzymes results in reduction in RNA fragment size over use of either alone.
  • Use RNase Cocktail for all situations, desirable to degrade RNA, i.e. plasmid minipreps and ribonuclease protection assays
  • RNase Cocktail is supplied in 50% glycerol for maximum convenience
  • Unit concentration: RNase A at 500U/mL and RNase T1 at 20,000U/mL
  • Quality Control: RNase Cocktail Enzyme Mix is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity; functionality is determined in ribonuclease protection assay
  • Unit Definitions: RNase A: One unit of RNase A is amount required to give increase in absorption at 286nm of 0.0146 absorbance units per minute in a 1mL volume
  • Unit assay conditions: 100mM Tris-acetate (pH 6.5), 1mM EDTA and 1mM cyclic 2', 3'-CMP
  • RNase T1: 100 Units of RNase T1 is amount of enzyme that yields an increase in absorption at 260nm of 0.01428 units per min. at room temperature using 60μg/mL yeast total RNA as a substrate
DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification
Order Info
Shipping Conditions: Dry ice
Specifications
Specifications
Dry Ice
RNase
RNAse Cocktail Enzyme Mix
Ambion™
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
For Research Use Only. Not for use in diagnostic procedures.