missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ RNase I, cloned, 100 U/μL DFS Item

Efficiently cleaves after all four bases of single-stranded RNA, in contrast to RNase A, which only cleaves after C and U residues

Supplier:  Invitrogen™ AM2294

 View more versions of this product

Catalog No. AM2294


Explore available promotions

Description

Description

RNase I degrades all RNA dinucleotide bonds leaving a 5' hydroxyl and 2', 3' cyclic monophosphate. RNase I will degrade any RNA to mixture of mononucleotides, dinucleotides, and trinucleotides and does not degrade DNA, although it will bind to DNA. It has marked preference for single-stranded RNA over double-stranded RNA, which allows it to work well in ribonuclease protection assays.

  • Supplied in one tube of 10,000U (100U/μL)
  • Endonuclease-free, exonuclease-free, protease-free
  • Rigorously tested for nonspecific contaminating endonuclease, exonuclease, and protease activity
  • Unit Definition: One unit is amount of enzyme required to produce 1μg of acid-soluble material from mouse liver RNA in 30 min. at 37°C

DNA & RNA Purification & Analysis, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting

Specifications

Specifications

100 U/μL
Dry Ice
10,000 U
Ambion™
-20°C
RNase
RNase I
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.