Invitrogen™ RT-PCR Grade Water DFS Item
RT-PCR Grade Water is certified free of nucleases, and free of nucleic acid contamination that may cause false-positive signals in RT-PCR. The RT-PCR Grade Water is ready to use and requires no preparation, mixing, or autoclaving.
Supplier:  Invitrogen™ AM9935
Catalog No. AM9935
Explore available promotions
Description
Description
RT-PCR Grade Water is certified free of nucleases, and free of nucleic acid contamination that may cause false-positive signals in RT-PCR. The RT-PCR Grade Water is ready to use and requires no preparation, mixing, or autoclaving.
Quality control
RT-PCR Grade Water is tested for prokaryotic as well as eukaryotic genomic DNA contamination using an ultrasensitive PCR assay. It is provided in 10 x 1.5 mL tubes. Prokaryotic DNA contamination is assessed by performing PCR with primers to 16S rRNA. Eukaryotic DNA contamination is assessed by performing PCR with primers to 18S rRNA. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity and is suitable for any PCR application.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
Molecular Biology
Not DEPC-Treated
Autoclaved, Membrane-Filtered
Ambion™
Tube
10 x 1.5 mL
Room Temperature
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.