Learn More
Invitrogen™ UltraPure™ BSA (50 mg/mL)
Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.
$194.00 - $770.00
Specifications
Content And Storage | Store at –20°C. |
---|---|
Concentration | 50 mg/mL |
Product Line | Ambion™ |
Shipping Condition | Dry Ice |
Form | Liquid |
Description
Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.
• Tubes containing 50 mg are provided, at a concentration of 50 mg/mL
• Bovine Serum Albumin (BSA) has many uses as a carrier protein and as a stabilizing agent in enzymatic reactions
• In northern, Southern, and dot blot hybridizations, BSA is also used as a blocking agent
• It is recommended in buffers for nick translation, polymerase reactions, and ligations
BSA is also a common additive for PCR amplifications, footprinting, and gel shift assays. In restriction digests, BSA has been shown to enhance enzyme activity. The molecular weight is 68 kD. Ultrapure BSA is rigorously tested for DNase, RNase, endonuclease, protease, peroxidase, and alkaline phosphatase activity, and assayed for fluorescence background. Exonuclease Activity: 3' - 5'.
Order Info
Shipping Condition: Dry Ice
Specifications
Store at –20°C. | |
Ambion™ | |
Liquid |
50 mg/mL | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.