missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Wash Solution I, RNA Purification
Total RNA Chemistry System component
Supplier: Invitrogen™ 4305891
Description
The protocol is designed for use with the 6100 Nucleic Acid PrepStation and 6700 Automated Nucleic Acid Workstation. This Wash Solution is for use with the Total RNA Chemistry System. Total RNA Chemistry makes possible the rapid isolation of total RNA from various samples, including tissue-cultured cells, whole blood, and peripheral blood mononuclear cells (PBMCs).
DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, RNA Extraction
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
6100 Nucleic Acid PrepStation, 6700 Automated Nucleic Acid Workstation | |
1000 mL | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.