American Scientific Bernoulli Bag
Helps explore Bernoulli's principle of air pressure.
Supplier: American Scientific 704000
Includes: Teacher's guide
Description
- Place it firmly to mouth (like balloon), it takes many breaths to blow it up
- When held in front of mouth and blown, air pressure in stream produce is reduced
- Entraps surrounding air to join in filling up bag, hence can be blown in single breath
- Effective after students have counted many of their own breaths in attempting to fill up bag
Specifications
Bernoulli Bag | |
Explores Bernoulli's principle of air pressure | |
Teacher's guide | |
4/Pk. |