Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System
Use for fast and easy detection of medium-to-high-target amounts. Cytiva Life Sciences™ Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction
Supplier: Cytiva RPN3006
Description
- Fast, easy detection of medium- to high-target amounts in 4 hr.
- Sufficient for hybridization of 4000cm2
Specifications
ECL™ Direct Nucleic Acid Labeling and Detection System | |
ECL Gold Hybridization Buffer sufficient for 4000 cm² | |
ECL Gold Hybridization Buffer | |
Sufficient for 4000 cm2 |