Cytiva Amersham™ Hyperfilm™ ECL
Use with all blue and green light chemiluminescent systems. Cytiva Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
Supplier: Cytiva 28906835
Description
- Antistatic coating reduces risk of visible artifacts
- Clear base offers high contrast for easy reading of images
- Suitable for use with all blue and green light chemiluminescent systems
- Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
- Chemiluminescent detection in Western, Southern, and Northern blots
Specifications
12.7 x 17.8 cm (5 x 7 in.) | |
Chemiluminescent detection in Western, Southern, and Northern blots. | |
4°C |
Amersham Hyperfilm ECL | |
50 Sheets/Box |