missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Amicon™ Stirred Cells

Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.

$1850.00 - $3110.00

Specifications

Autoclavable Autoclavable
For Use With (Equipment) Ultracel™, Biomax™, and Durapore™ membrane discs for use in stirred cells
For Use With (Application) Concentration, Buffer Exchange, Desalting
Type Ultrafiltration Equipment
View More Specs

Alert:

Pressure Limitation. Do not operate the stirred cell above 5.2 bar (75 psi).


Products 3
Catalog Number Mfr. No. Volume (Metric) Hold Up Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Hold Up Price Quantity  
UFSC05001
View Documents
MilliporeSigma™
UFSC05001
50 mL
Each for $1,850.00
 
UFSC20001
missing translation for 'viewDocuments'
MilliporeSigma™
UFSC20001
200 mL
Each for $2,340.00
 
UFSC40001
missing translation for 'viewDocuments'
MilliporeSigma™
UFSC40001
400 mL
Each for $3,110.00
 
Description

Description

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.
Specifications

Specifications

Autoclavable
Ultracel™, Biomax™, and Durapore™ membrane discs for use in stirred cells
Concentration, Buffer Exchange, Desalting
Ultrafiltration Equipment
Videos
SDS
Documents

Documents