missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amido Black 10B, Spectrum™ Chemical
$215.59 - $2427.15
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18608566
|
Spectrum Chemical Mfg Cor
AM11025GM |
25 g | Amber Glass Bottle |
Each for $215.59
|
|
|||||
18608565
|
Spectrum Chemical Mfg Cor
AM110100GM |
100 g | Amber Glass Bottle |
Each for $467.35
|
|
|||||
18608564
|
Spectrum Chemical Mfg Cor
AM1101KG |
1 kg | Amber Glass Bottle |
Each for $2,427.15
|
|
|||||
Chemical Identifiers
1064-48-8 |
Specifications
1064-48-8 | |
Reagent | |
Dark Brown | |
Amido Black 10B |
1 | |
Amber Glass Bottle | |
25 g |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.