missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amido Black 10B, Thermo Scientific Chemicals

Used as a histological, protein, and nucleic acid dye used in electrophoresis.

Supplier:  Thermo Scientific Chemicals A1137414

 View more versions of this product

Catalog No. AAA1137414


Explore available promotions

Description

Description

Amido Black 10B is used as a histological, protein, and nucleic acid dye used in electrophoresis. Naphthol Blue Black is an important and commonly used industrial dye. This dye is also used in rapid staining of protein bands on nitrocellulose membranes such as Western blots. Naphthol Blue Black can be utilized as a nucleic acid stain in SDS-Page gels. Additionally this compound has been untilized in cell proliferation assays and histochemical studies. It is also used in criminal investigations to detect blood present with latent fingerprints. They are also used in paints, inks, plastics and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Amido Black 10B
C22H14N6Na2O9S2
5374263
It is soluble in water (10mg/ml at 25°C),and ethanol.
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
616.487
616.49
Hygroscopic
Powder
1064-48-8
MFCD00004017
C.I. 20470,Naphthalene Black 10B
HKBVRFLHNUEVRO-UHFFFAOYSA-L
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
44134531
Glass bottle
25 g

Chemical Identifiers

1064-48-8
616.487
HKBVRFLHNUEVRO-UHFFFAOYSA-L
44134531
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C22H14N6Na2O9S2
MFCD00004017
C.I. 20470,Naphthalene Black 10B
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P314-P332+P313-P362-P501c

H315-H317-H319-H335-H372-H500

EINECSNumber : 213-903-1

RTECSNumber : QJ6196000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only