missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminoacetaldehyde dimethyl acetal, 99%, Thermo Scientific Chemicals

$101.36 - $942.30

Chemical Identifiers

CAS 22483-09-6
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00008135
InChI Key QKWWDTYDYOFRJL-UHFFFAOYSA-N
Synonym aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr
PubChem CID 89728
IUPAC Name 2,2-dimethoxyethanamine
SMILES COC(CN)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1549814
View Documents
Thermo Scientific Chemicals
A1549814
25 g N/A
 
AAA1549822
View Documents
Thermo Scientific Chemicals
A1549822
100 g N/A
 
AAA1549836
View Documents
Thermo Scientific Chemicals
A1549836
500 g N/A
 
Description

Description

Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

Solubility
Miscible in water. Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications

Chemical Identifiers

22483-09-6
105.137
QKWWDTYDYOFRJL-UHFFFAOYSA-N
89728
COC(CN)OC
C4H11NO2
MFCD00008135
aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr
2,2-dimethoxyethanamine

Specifications

22483-09-6
135°C to 138°C
C4H11NO2
MFCD00008135
UN2733
aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr
QKWWDTYDYOFRJL-UHFFFAOYSA-N
2,2-dimethoxyethanamine
89728
99%
0.973
41°C (106°F)
1.417
25 g
741868
Miscible in water. Soluble in chloroform,methanol.
COC(CN)OC
105.137
105.14
Aminoacetaldehyde dimethyl acetal
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 245-026-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only