missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aminoacetonitrile hydrochloride, 98+%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 04198414

Catalog No. AA4198414


Explore available promotions

Description

Description

Used as pharmaceutical intermediates, organic synthetic raw material transportation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aminoacetonitrile hydrochloride
6011-14-9
25 g
C2H5ClN2
aminoacetonitrile hydrochloride, 2-aminoacetonitrile hydrochloride, glycinonitrile hydrochloride, aminoacetonitrile hcl, unii-s8l3v1ksb7, acetonitrile, amino-, monohydrochloride, s8l3v1ksb7, aminoacetonitrile.hcl, amino-acetonitrile hcl, acmc-209mhz
XFKYKTBPRBZDFG-UHFFFAOYSA-N
2-aminoacetonitrile;hydrochloride
11491918
Hygroscopic
166°C to 171°C (decomposition)
Powder
>98%
MFCD00012850
Soluble in water 20°C,1000g/L.
C(C#N)N.Cl
92.526
92.53

Chemical Identifiers

6011-14-9
92.526
XFKYKTBPRBZDFG-UHFFFAOYSA-N
11491918
C(C#N)N.Cl
C2H5ClN2
MFCD00012850
aminoacetonitrile hydrochloride, 2-aminoacetonitrile hydrochloride, glycinonitrile hydrochloride, aminoacetonitrile hcl, unii-s8l3v1ksb7, acetonitrile, amino-, monohydrochloride, s8l3v1ksb7, aminoacetonitrile.hcl, amino-acetonitrile hcl, acmc-209mhz
2-aminoacetonitrile;hydrochloride
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 227-865-9

RTECSNumber : MC1950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only