Learn More
Aminoguanidine bicarbonate, 98.5%
CAS: 2582-30-1 | CH6N4·H2CO3 | 136.11 g/mol
Supplier: Thermo Scientific Chemicals 149060010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Aminoguanidine bicarbonate | |
| 125°C | |
| 0.3% max. | |
| 98.5% | |
| CH6N4·H2CO3 | |
| 1 kg | |
| aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate | |
| OTXHZHQQWQTQMW-UHFFFAOYSA-N | |
| 2-aminoguanidine;carbonic acid | |
| 164944 | |
| 98.50% |
| 2582-30-1 | |
| White | |
| Authentic | |
| Plastic Bottle | |
| H2NNHC(=NH)NH2·H2CO3 | |
| 03,117 | |
| Solubility in water: <5g/L (20°C) | |
| C(=NN)(N)N.C(=O)(O)O | |
| 136.11 | |
| 136.11 | |
| Crystalline Powder |
Chemical Identifiers
| 2582-30-1 | |
| 136.11 | |
| aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate | |
| 2-aminoguanidine;carbonic acid |
| CH6N4·H2CO3 | |
| OTXHZHQQWQTQMW-UHFFFAOYSA-N | |
| 164944 | |
| C(=NN)(N)N.C(=O)(O)O |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May damage fertility or the unborn child.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention
GHS Signal Word: Danger
EINECSNumber : 219-956-7
RUO – Research Use Only