missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine hydrochloride, 98%

CAS: 1937-19-5 | CH6N4·HCl | 110.55 g/mol

$108.47 - $108.47

Chemical Identifiers

CAS 1937-19-5
Molecular Formula CH6N4·HCl
Molecular Weight (g/mol) 110.55
MDL Number MFCD00039074
InChI Key UBDZFAGVPPMTIT-UHFFFAOYSA-N
Synonym aminoguanidine hydrochloride, pimagedine hcl, guanylhydrazine hydrochloride, hydrazinecarboximidamide hydrochloride, aminoguanidinehydrochloride, aminoguanidine hcl, pimagedine hydrochloride, aminoguanidine monohydrochloride, hydrazinecarboximidamide, hydrochloride
PubChem CID 2734687
IUPAC Name 2-aminoguanidine;hydrochloride
SMILES C(=NN)(N)N.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368910250
SDS View Documents
Thermo Scientific Chemicals
368910250
25 g Glass bottle
Each for $108.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1937-19-5
110.55
UBDZFAGVPPMTIT-UHFFFAOYSA-N
2734687
C(=NN)(N)N.Cl
CH6N4·HCl
MFCD00039074
aminoguanidine hydrochloride, pimagedine hcl, guanylhydrazine hydrochloride, hydrazinecarboximidamide hydrochloride, aminoguanidinehydrochloride, aminoguanidine hcl, pimagedine hydrochloride, aminoguanidine monohydrochloride, hydrazinecarboximidamide, hydrochloride
2-aminoguanidine;hydrochloride

Specifications

1937-19-5
White
98%
CH6N4·HCl
MFCD00039074
03, IV, 236
aminoguanidine hydrochloride, pimagedine hcl, guanylhydrazine hydrochloride, hydrazinecarboximidamide hydrochloride, aminoguanidinehydrochloride, aminoguanidine hcl, pimagedine hydrochloride, aminoguanidine monohydrochloride, hydrazinecarboximidamide, hydrochloride
UBDZFAGVPPMTIT-UHFFFAOYSA-N
2-aminoguanidine;hydrochloride
2734687
98%
Aminoguanidine hydrochloride, 98%
>160.0°C
Authentic
Glass bottle
H2NNHC(=NH)NH2·HCl
25 g
15, 438
Solubility in water: soluble.
C(=NN)(N)N.Cl
110.55
110.55
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-707-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only