missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(Aminomethyl)phosphonic Acid, TCI America™
$271.62 - $802.76
Chemical Identifiers
CAS | 1066-51-9 |
---|---|
Molecular Formula | CH6NO3P |
Molecular Weight (g/mol) | 111.037 |
MDL Number | MFCD00008105 |
InChI Key | MGRVRXRGTBOSHW-UHFFFAOYSA-N |
Synonym | aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl |
PubChem CID | 14017 |
ChEBI | CHEBI:28812 |
IUPAC Name | aminomethylphosphonic acid |
SMILES | C(N)P(=O)(O)O |
Chemical Identifiers
1066-51-9 | |
111.037 | |
MGRVRXRGTBOSHW-UHFFFAOYSA-N | |
14017 | |
aminomethylphosphonic acid |
CH6NO3P | |
MFCD00008105 | |
aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl | |
CHEBI:28812 | |
C(N)P(=O)(O)O |
Specifications
1066-51-9 | |
CH6NO3P | |
1 g | |
MGRVRXRGTBOSHW-UHFFFAOYSA-N | |
aminomethylphosphonic acid | |
14017 | |
111.04 | |
Crystalline Powder |
White | |
MFCD00008105 | |
aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl | |
C(N)P(=O)(O)O | |
111.037 | |
CHEBI:28812 | |
≥97.0% (T) | |
(Aminomethyl)phosphonic Acid |
Safety and Handling
RTECSNumber : SZ6526000
TSCA : No