Brady™ Ammonia (IIAR) Pipe Markers, Legend: LSS/VAP/Ammonia/LOW
For use in labeling ammonia piping and its contents. Brady Ammonia (IIAR) Pipe Markers help create a safer, more efficient work environment.
Supplier:  Brady™ 57993
 View more versions of this product
Catalog No. 19-110-485
Explore available promotions
Description
Description
  • Meets IIAR Standard and is recommended for use in labeling ammonia piping and its contents
  • Made of B-689 high performance wrap-around PVF over-laminated polyester and B-681/B-883 polyester over-laminate on fiberglass plastic carrier
Specifications
Specifications
Style II
PVF Over-laminated Polyester (B-689)
IIAR Abbreviation: LSS; Physical State: VAP; Pressure Level: LOW; Legend: Ammonia
12 x 26 in. (W x H)
1.25 in.
2 to 8 in.
Black, White, Sky Blue, Green, Yellow
Ammonia (IIAR) Pipe Marker
Pipe and valve marking
304.8 x 660.4 mm (W x H)
31.75 mm
50.8 to 203.2 mm