missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Acetate, ∽99%, (Crystalline), MP Biomedicals
$422.91 - $706.48
Chemical Identifiers
CAS | 631-61-8 |
---|---|
Molecular Formula | C2H7NO2 |
Molecular Weight (g/mol) | 77.083 |
InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
Synonym | ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 |
PubChem CID | 517165 |
ChEBI | CHEBI:62947 |
IUPAC Name | azanium;acetate |
SMILES | CC(=O)[O-].[NH4+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
ICN10065102
|
MP Biomedicals Inc
0210065102 |
2 kg |
Each for $422.91
|
|
||||||
ICN10065105
|
MP Biomedicals Inc
0210065105 |
5 kg |
Each for $706.48
|
|
||||||
Chemical Identifiers
631-61-8 | |
77.083 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CHEBI:62947 | |
CC(=O)[O-].[NH4+] |
C2H7NO2 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
517165 | |
azanium;acetate |
Specifications
631-61-8 | |
2 kg | |
C2H7NO2 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
azanium;acetate | |
517165 | |
77.1 |
Crystalline | |
∽99% | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CC(=O)[O-].[NH4+] | |
77.083 | |
CHEBI:62947 | |
Ammonium Acetate |
Safety and Handling
Recommended Storage : Room Temperature