missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Acetate (Crystalline/Certified ACS), Fisher Chemical™ Edge
Edge

Supplier:  Fisher Chemical FLA637500

 View more versions of this product

Catalog No. A637-500



Edge

Description

Description

Specifications

Specifications

Ammonium Acetate
631-61-8
7.0
500 g
C2H7NO2
MFCD00013066
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
≥97%
Pass Test
1.07g/mL
114°C
White
Solid
≥97%
CH3COONH4
136°C
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
77.08
Certified ACS
Poly Bottle

Chemical Identifiers

631-61-8
77.083
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate
C2H7NO2
MFCD00013066
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:1

SDS Documents

Documents

Promotions

Promotions