missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Acetate, For Analysis, EMSURE™, MilliporeSigma™

Supplier:  Technidata 1011161000

 View more versions of this product

Catalog No. M1011161000


Explore available promotions

Description

Description

Recommended Storage

+15°C to +25°C.

Specifications

Specifications

114°C
631-61-8
Solid
C2H7NO2
1480g/L
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
Plastic Bottle
Colorless
6.0 to 7.5
1 kg
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
>96%
1.17 g/3 (20 °C)

Chemical Identifiers

631-61-8
77.083
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate
Safety and Handling

Safety and Handling

EINECSNumber : 211-162-9

RTECSNumber : AF3675000

Recommended Storage : +15°C to +25°C.

SDS
Documents

Documents

Promotions

Promotions