missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bicarbonate, 98%, pure

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol

$73.17 - $254.95

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC370930250
SDS View Documents
Thermo Scientific Chemicals
370930250
25 g Glass bottle
Each for $73.17
Only null left
 
AC370930010
SDS View Documents
Thermo Scientific Chemicals
370930010
1 kg Glass bottle
Each for $163.30
Only null left
 
AC370930050
SDS View Documents
Thermo Scientific Chemicals
370930050
5 kg Plastic Drum
Each for $254.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1066-33-7
79.06
ATRRKUHOCOJYRX-UHFFFAOYSA-N
14013
N.OC(O)=O
CH5NO3
MFCD00012138
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
carbonic acid amine

Specifications

105°C
White
25 g
CH5NO3
MFCD00012138
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
ATRRKUHOCOJYRX-UHFFFAOYSA-N
carbonic acid amine
14013
98%
Authentic
Ammonium bicarbonate
1066-33-7
Crystalline Powder, Crystals and/or Chunks
98%
NH4HCO3
15,494
Solubility in water: 220g/L (20°C)
N.OC(O)=O
79.06
79.06
Pure
Glass bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

GHS Signal Word: Warning

EINECSNumber : 213-911-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only