missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Bifluoride (Crystalline/Technical), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLA664500

 View more versions of this product

Catalog No. A664-500

Explore available promotions

Description

Description

Specifications

Specifications

230°C
125°C
1341-49-7
Solid
F2H5N
Ammonium Acid Fluoride, Ammonium Hydrogen Difluoride
N.F.F
57.04
Technical
Poly Bottle
>230°C
Ammonium Bifluoride
White
3.5
500 g
MFCD00012018
KVBCYCWRDBDGBG-UHFFFAOYSA-N
amine dihydrofluoride
57.04
Pass Test
1hPa at 20°C
1.50g/cm³

Chemical Identifiers

1341-49-7
57.04
KVBCYCWRDBDGBG-UHFFFAOYSA-N
amine dihydrofluoride
F2H5N
MFCD00012018
Ammonium Acid Fluoride, Ammonium Hydrogen Difluoride
N.F.F
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions